Learn More
Fine Science Tools Vannas Spring Scissors - 3mm Cutting Edge, Curved
Catalog No. NC9091940
Encompass
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 3mm/ Tip Diameter: 0.05mm/ Alloy / Material: Stainless Steel/ Length: 8cm/ Serrated: No. These scissors have been designed to maximize visibility under the microscope by eliminating unnecessary bulk in the blades, and they have a long taper which is ideal for cutting vessel walls.
Catalog No. NC9091940 Supplier Fine Science Tools Supplier No. 1500010
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 3mm/ Tip Diameter: 0.05mm/ Alloy / Material: Stainless Steel/ Length: 8cm/ Serrated: No. These scissors have been designed to maximize visibility under the microscope by eliminating unnecessary bulk in the blades, and they have a long taper which is ideal for cutting vessel walls.