Learn More
Fine Science Tools Spring Scissors, 8mm Cutting Edge, Stainless Steel, 8.5 cm
Catalog No. NC0127560
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 8mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm/ Serrated: No
Catalog No. NC0127560 Supplier Fine Science Tools Supplier No. 1500908
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 8mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm/ Serrated: No