missing translation for 'onlineSavingsMsg'
Learn More

STERIS Spor-Klenz™ Ready-To-Use Sterilant/Disinfectant

Provides consistent microbial control, no mixing or activation is required. Spray or apply manually straight out of the container.

$108.03

Specifications

Product Type Cleanroom Disinfectant, Sterilant, and Sporicidal Cleaner
Certifications/Compliance European Union Biocidal Products Directive (BPD)
Filtered Filtered
For Use With (Application) Sterilization and Disinfection of Hard Environmental Surfaces
Quantity 4/Cs.
View More Specs

Products 1
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
0432420
View Documents Promotion Details
STERIS
652501CA
3200 mL
Each for $108.03
Case of 4 Each for $432.12
Only null left
 
Description

Description

  • Specifically formulated for the sterilization and disinfection of hard environmental surfaces
  • A stabilized blend of hydrogen peroxide, peracetic, and acetic acids
  • Provides effective microbial control, including spores
  • Filtered to 0.22 μm
  • Offers a low toxicity profile that requires no mixing or activation
  • Reusable for up to 14 days to reduce waste
  • For use in Pharmaceutical, Medical Device, Biotech, and Cosmetic manufacturing facilities
  • All of the active ingredients are notified according to the European Union Biocidal Products Directive (BPD)
  • Each lot is fully traceable and documented by a Certificate of Analysis including filtration and USP sterility
  • Note: Sprayer is sold separately (see Cat No. 6526Q8)   
Specifications

Specifications

Cleanroom Disinfectant, Sterilant, and Sporicidal Cleaner
Filtered
4/Cs.
European Union Biocidal Products Directive (BPD)
Sterilization and Disinfection of Hard Environmental Surfaces
Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

SDS
Documents

Documents