missing translation for 'onlineSavingsMsg'
Learn More

STERIS Spor-Klenz™ Ready To Use Cold Sterilant, ETO Processed Packaged

Catalog No. 19415150
Change view
Click to view available options
Volume (English):
1 qt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19415150 1 qt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19415150 Supplier STERIS Supplier No. 652873CA
Only null left

A fast-acting formula that provides microbial control against a broad spectrum of organisms, and can easily be introduced into cleanroom environments

  • A liquid sporicide specifically formulated for the sterilization and disinfection of hard environmental surfaces
  • Stabilized blend of peracetic acid, hydrogen peroxide, and acetic acid
  • Microbial control against bacteria, fungi, viruses, and bacteria spores proven using AOAC and EN protocols
  • Can be applied manually, sprayed, or fogged straight out of the container
  • Product is ETO processed through a validated process and double bagged for easy introduction into aseptic areas
  • For use in Pharmaceutical, Medical Device, Biotech, and Cosmetic manufacturing facilities
  • Requires no mixing or activation
  • Reusable for up to 14 days to reduce waste
  • All of the active ingredients are notified according to the European Union Biocidal Products Directive (BPD)
  • Each lot is fully traceable and documented by a Certificate of Analysis that includes filtration and USP sterility

Specifications

Product Type Cold Sterilant Disinfectant
Sterility Sterile
Composition Stabilized Blend of Peracetic Acid, Hydrogen Peroxide, and Acetic Acid
Container Type Trigger Spray Bottles
For Use With (Application) Sterilizing and Disinfecting Hard Environmental Surfaces
Quantity 6/Cs.
Volume (English) 1 qt.
Volume (Metric) 946 mL