Learn More
MilliporeSigma™ Supelco™ SPME-OC Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)
Catalog No. 111008084
Change view
Click to view available options
Quantity:
3/Pk.
Catalog No. Quantity
111008084 3/Pk.
1 options
Catalog No. 111008084 Supplier MilliporeSigma™ Supelco™ Supplier No. 57439U
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description df 75 μm (Coating thickness includes 65 μm coating + 10 μm OC (overcoating)), needle size 23 ga, 3/Pk.)
Material Notched Pink Hub
Quantity 3/Pk.