Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Divinylbenzene/Carboxen™/Polydimethylsiloxane (DVB/CAR/PDMS), df 50/30 μm
Catalog No. p-7194116
Change view
Click to view available options
Needle Gauge:
23 ga, L: 1 cm
23 ga; L: 2 cm
23 ga
24 ga
For Use With (Equipment):
Autosampler
Manual Holder
Description:
Notched Gray Hub
Plain Gray Hub
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge For Use With (Equipment) Description
111007791 23 ga, L: 1 cm Autosampler -
111007792 23 ga; L: 2 cm Autosampler -
111020745 23 ga Autosampler -
111020754 24 ga Autosampler -
111020746 23 ga Manual Holder -
111020755 24 ga Manual Holder Plain Gray Hub
111020756 24 ga Manual Holder Notched Gray Hub
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 111007791 Supplier MilliporeSigma™ Supelco™ Supplier No. 57912U
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS) is manufactured with a flexible metal alloy used in the needle, plunger, and fiber core.
The new metal alloy design includes a thicker, flexible plunger that is much less likely to kink or break, and helps to reinforce the needle especially when used in an auto-sampler with a sample agitator. Since the needle is more flexible and has a thinner wall than the standard stainless steel needle, a bevel has been placed on the needle to help it pierce septa materials more easily.
Specifications
For Use With (Application) SPME
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material Metal Alloy Fiber
Needle Gauge 23 ga, L: 1 cm
Quantity 1/Pk.
Carboxen and StableFlex are registered trademarks of Sigma-Aldrich Co. LLC