Learn More
Splash Shield™ Face Shields
Catalog No. 17987214
Change view
Click to view available options
Product Type:
Long Faceshield
Short Faceshield
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17987214 Short Faceshield
17987213 Long Faceshield
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17987214 Supplier Splash Shield™ Supplier No. 4541S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
This is the replacement shield only; frame is sold separately.
Lightweight, comfortable and cost-efficient.
Maximum face protection for hospitals, dental offices, labs and EMT use.
  • 33.02cm (13 in.) impenetrable foam barrier for wide coverage
  • Lightweight with no head strap
  • Optically clear, anti-fog and latex-free
Specifications
Lens Coating Anti-fog
Lens Tint Clear
Product Type Short Faceshield
For Use With (Application) Maximum face protection for hospitals, dental offices, labs and EMT use
Length (English) 5.75 in.
Length (Metric) 14.6 cm
No. per Case 200/Cs.