missing translation for 'onlineSavingsMsg'
Learn More

Spironolactone

Catalog No. p-7041752
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6011903 1 g
AAJ6011906 5 g
AAJ6011909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6011903 Supplier Thermo Scientific Chemicals Supplier No. J6011903
Only null left

CAS: 52-01-7 | C24H32O4S | 416.576 g/mol

Spironolactone used as potassium sparing diuretic. Also used primarily to treat ascites in patients with liver disease, low-renin hypertension, hypokalemia, and Conn’s syndrome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform and methanol. Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 52-01-7
Molecular Formula C24H32O4S
Molecular Weight (g/mol) 416.576
MDL Number MFCD00082250
InChI Key LXMSZDCAJNLERA-ZHYRCANASA-N
PubChem CID 5833
ChEBI CHEBI:9241
IUPAC Name S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
SMILES CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C

Specifications

Type Spironolactone
Physical Form Powder
Melting Point 207°C to 208°C
Color White
Quantity 1 g
Merck Index 14,8760
Solubility Information Soluble in chloroform and methanol. Slightly soluble in water.
Formula Weight 416.58
Chemical Name or Material Spironolactone
Hazard Category H351-H360F-H373
Hazard Statement GHS H Statement
H360-H302
May damage fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 200-133-6
RTECSNumber TU4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only