missing translation for 'onlineSavingsMsg'
Learn More

Spironolactone 98.0+%, TCI America™

Catalog No. p-7135850
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02601G 1 g
S02605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S02601G Supplier TCI America Supplier No. S02601G
Only null left

Chemical Identifiers

CAS 52-01-7
Molecular Formula C24H32O4S
Molecular Weight (g/mol) 416.576
MDL Number MFCD00082250
InChI Key LXMSZDCAJNLERA-ZHYRCANASA-N
Synonym spironolactone, aldactone, spirolactone, verospiron, euteberol, spiroctan, spirolang, verospirone, aldactone a, spironocompren
PubChem CID 5833
ChEBI CHEBI:9241
IUPAC Name S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
SMILES CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C

Specifications

Color White
Quantity 1 g
Formula Weight 416.58
Percent Purity ≥95.0% (E)
Physical Form Crystalline Powder
Chemical Name or Material Spironolactone
RTECSNumber TU4725000
TSCA Yes