Spindle guard made of stainless steel AISI 316L (1.4404 / 1.4435). Comes with a fixing screw with identification """"L"""". The screw also serves as guard detection, which avoids measurement with a wrong configuration (TruGuard). For use inside wide-rimmed vessels (typically 600 mL glass beakers). Required for spindles L1 and L2.Measurement Spindles L1 and L2 included with ViscoQCSuitable for use with Anton Paar rotational viscometers ViscoQC 100 L item NC1818559, ViscoQC 300 L item NC1818572.