Learn More
Can-Am Instruments Ltd SPINDLE DISPOSABLE 100/PK
Catalog No. NC9374343
Encompass
Disposable aluminum spindle SC4-27D Pack of 100 for Small Sample Adapter or Thermosel accessory
Catalog No. NC9374343 Supplier Can-Am Instruments Ltd Supplier No. SC427D100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Disposable aluminum spindle SC4-27D Pack of 100 for Small Sample Adapter or Thermosel accessory