missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ SpillStopper™ Work Surfaces Cupsink Cover

Catalog No. 16317158
Change view
Click to view available options
For Use With (Equipment):
SpillStopper™ Work Surfaces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
16317158 SpillStopper™ Work Surfaces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16317158 Supplier Labconco™ Supplier No. 4005201

SpillStopper™ Cupsink Cover provides a continuous work surface when a cupsink is not in use

Black, solid epoxy resin cover for oval cupsink cutout

Specifications

Color Black
Material Epoxy
Type Cupsink Cover
For Use With (Equipment) SpillStopper™ Work Surfaces
Dimensions (L x W x H) 4.4 x 7.4 x 0.3 in.
Weight (English) 1.5 lb.
Weight (Metric) 0.7 kg