missing translation for 'onlineSavingsMsg'
Learn More

FyterTech Nonwovens Spilfyter™ Wastewater Classifiers

Catalog No. 19812534
Change view
Click to view available options
Type:
Wastewater Classifier Kit
Wastewater Classifier Test Strip
Packaging Quantity:
10/Pk.
50/Pk.
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Packaging Quantity
19812534 Wastewater Classifier Test Strip 50/Pk.
19065574 Wastewater Classifier Test Strip 10/Pk.
19120875 Wastewater Classifier Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19812534 Supplier FyterTech Nonwovens Supplier No. 580050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Distinguish risks of unknown spilled liquids

Classifier Strips show responder if unknown spill contains critical levels of these contaminant categories: acid/base; petroleum/organic solvent; hydrogen sulfide; nitrite; nitrate; fluoride. Classifier chart compares standard colors to sample colors.

Wastewater Classifier Kit consists of:

  • Six Wastewater Classifier Strips
  • One bobbin of polypropylene thread
  • Laminated Chemical Classifier Chart
Wastewater Classifier Station consists of:
  • 20 Wastewater Classifier Strips
  • Two bobbins of polypropylene thread
  • Laminated Chemical Classifier Chart
  • One pair nitrile gloves
  • One plastic case

Order Info

Additional test strips and classifier charts are available separately.

Specifications

Description 20 ppm fluoride, 10 ppm petroleum products/organic solvents, 10 ppm hydrogen sulfide, 10 ppm nitrates, 1 ppm nitrites
Type Wastewater Classifier Test Strip
For Use With (Application) Classifying the wastewater for proper treatment
Includes 50 strips
Packaging Quantity 50/Pk.
pH Range 0 to 13