missing translation for 'onlineSavingsMsg'
Learn More

FyterTech Nonwovens Spilfyter™ pH Paper

Catalog No. 19120829
Change view
Click to view available options
pH Range:
0 to 13
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
19120829 0 to 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19120829 Supplier FyterTech Nonwovens Supplier No. RCC7018
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Wide-range, general-purpose pH test strips

  • Determines pH from 0 to 13
  • 14 brilliant colors
  • 0.25 in.W × 10 ft.L (1.3 × 305cm)
  • Packaged in convenient dispenser with color comparison chart on side

Specifications

Type Spilfyter pH Paper
For Use With (Application) General purpose and determines pH from 0 to 13
Includes 14 Brilliant Colors comparison chart
Packaging Quantity 1 Roll
pH Range 0 to 13