Learn More
FyterTech Nonwovens Spilfyter™ KolorSafe™ Liquid Neutralizers for Bases
Take the guesswork out of neutralizing hazardous spills
Supplier:  FyterTech Nonwovens JV430001
 View more versions of this product
Catalog No. 19-021890
Only null left
Description
Description
Unlike dry neutralizer, liquid reagent can be sprayed into hard-to-reach areas and on vertical surfaces.
  • Sodium-free liquid reagent changes color when neutralization is complete
  • Safe for use in laboratories and cleanrooms
Specifications
Specifications
0.24 gal.
Base Neutralizer
For lab and cleanrooms
12/Cs.
14 kg
0.94 L
Liquid
Color change indicator when neutralized
31 lb.
Product Suggestions
Product Suggestions