missing translation for 'onlineSavingsMsg'
Learn More

FyterTech Nonwovens Spilfyter™ KolorSafe™ Neutralizers for Acids

Take the guesswork out of neutralizing hazardous spills

Supplier:  FyterTech Nonwovens JV 440001

 View more versions of this product

Catalog No. 19021893


Only null left

Description

Description

 

  • Sodium-free liquid reagent changes color when neutralization is complete
  • Safe for use in laboratories and cleanrooms

 

Specifications

Chemical Identifiers

Spill control
Color change indicator when neutralized

Specifications

2 lb.
Dry Acid Neutralizer
Spill control
10/Cs.
10 kg
0.9 kg
Dry Acid Neutralizer
Color change indicator when neutralized
23 lb.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 5 Years

SDS
Documents

Documents

Promotions

Promotions