missing translation for 'onlineSavingsMsg'
Learn More

FyterTech Nonwovens Spilfyter™ KolorSafe™ Neutralizers for Acids

Catalog No. 19021888
Change view
Click to view available options
Type:
Acid Neutralizer
Dry Acid Neutralizer
Liquid Acid Neutralizer
Capacity (Metric):
0.9 kg
0.94 L
3.78 L
18.92 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Capacity (Metric)
19021888 Acid Neutralizer 0.94 L
19066518 Liquid Acid Neutralizer 18.92 L
19021889 Acid Neutralizer 3.78 L
19021893 Dry Acid Neutralizer 0.9 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19021888 Supplier FyterTech Nonwovens Supplier No. JV 410001

Take the guesswork out of neutralizing hazardous spills

Unlike dry neutralizer, liquid reagent can be sprayed into hard-to-reach areas and on vertical surfaces.

  • Sodium-free liquid reagent changes color when neutralization is complete
  • Safe for use in laboratories and cleanrooms

Specifications

Capacity (English) 0.24 gal.
Capacity (Metric) 0.94 L
Type Acid Neutralizer
Format Liquid
For Use With (Application) Spill control
Includes Color change indicator when neutralized
Packaging Quantity 12/Cs.
Weight (English) 31 lb.
Weight (Metric) 14 kg