Learn More
Justrite™ Spigot Fitting
GREENER_CHOICE
Catalog No. 19830502
Change view
Click to view available options
Closure Type:
1-1/8 in. Thread with 1/2 in. Hose Barb
Catalog No. Closure Type
19830502 1-1/8 in. Thread with 1/2 in. Hose Barb
1 options
Catalog No. 19830502 Supplier Justrite™ Supplier No. 12904
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For leakproof, controlled fluid transfer.
  • With easy-grip lever to control continuous or manual flow rates
  • Designed to withstand 100L pressure
  • With closed cap
Specifications
Quantity Each
Product Type Spigot Fitting
For Use With (Equipment) Carboys
Material Polypropylene
Closure Type 1-1/8 in. Thread with 1/2 in. Hose Barb