missing translation for 'onlineSavingsMsg'
Learn More

Beckman Coulter Spigot Assembly

Utilize the Spigot Assembly for easy dispensing of ISOTON II and Coulter CLENZ Cleaning Agents.

Supplier:  Beckman Coulter 8531425

Catalog No. 15655121


Only null left

Description

Description

  • Aids in dispensing of ISOTON II (20 L), and Coulter Clenz (5 L)
  • Easy to install
Specifications

Specifications

Spigot Assembly
ISOTON II (20 L) and Coulter Clenz (5 L)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions