Learn More
SignalChem Biotech PI4KA, Active 50ug
Catalog No. 502573769
Encompass
Recombinant full-length human PI4KA was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502573769 Supplier SignalChem Biotech Supplier No. P31102G50
Recombinant full-length human PI4KA was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.