Learn More
Cole-Parmer™ Hydraulic Laboratory Pellet Press Accessories, Die Sleeve-and-plunger Set
Optimize performance of your Cole-Parmer™ hydraulic laboratory pellet press with this Die Sleeve-and-plunger Set.
Supplier: Cole-Parmer™ 3616W
Description
- Choose from a wide range of accessories including pellet die sets, pellets, flaring tool, and more.
- Ensure top results from your Cole-Parmer™ hydraulic laboratory pellet press for pressing of sample pellets for XRF and other analytical techniques with these accessories and components.
- The accessories and replacement parts include the 13 mm, 31 mm, 35 mm, and 40 mm pellet die sets, different material pellets, sleeve-and plunger sets, and more.
Chemical Identifiers
| Sleeve And Plunger Set | |
| Instructions |
Specifications
| Sleeve And Plunger Set | |
| 35 mm | |
| For 3616 35 mm Evacuable Pellet Die Set and Any Prepaid™ Sample Binder |
| 1.3 in. | |
| For Preparation of XRF Disks with Binder Matrix Supporting a Layer of Sample | |
| Instructions |