Learn More
Cole-Parmer™ Hydraulic Laboratory Pellet Press Accessories, Die Set Pellet
Optimize performance of your Cole-Parmer™ hydraulic laboratory pellet press with this Die Set Pellet.
Supplier:  Cole-Parmer™ SPX3614
 View more versions of this product
Catalog No. 3614
Only null left
Description
Description
  • Choose from a wide range of accessories including pellet die sets, pellets, flaring tool, and more.
  • Ensure top results from your Cole-Parmer™ hydraulic laboratory pellet press for pressing of sample pellets for XRF and other analytical techniques with these accessories and components.
  • The accessories and replacement parts include the 13 mm, 31 mm, 35 mm, and 40 mm pellet die sets, different material pellets, sleeve-and plunger sets, and more.
Specifications
Chemical Identifiers
Die Set Pellet
Specifications
Die Set Pellet
40 mm
For XRF Analysis, 3616 W 35 mm Sleeve and Plunger Set
10 lb.
1.5 in.
To Form 40 mm KBR Disks
Evacuable die body, base, plunger, two 40 mm polished steel pellets, o-ring vacuum seals and knock out ring for sample disk extraction
4.5 kg
Product Suggestions
Product Suggestions