Learn More
Cole-Parmer™ XRF Prep-Aid™ Grinding Aids and Sample Binders
Prepare samples for reproducible and contamination-free results with XRF Prep-Aid™ Grinding Aids and Sample Binders.
Supplier: Cole-Parmer™ 3646150
Description
Ideal for samples which crumble or ablate after pressing.
- Select binders from powder, tablet, or liquid form depending on your application.
- High-quality sample preparation is the key to achieving consistent and reliable analytical results.
- Powders are ideal for samples that crumble or ablate after pressing.
- They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks.
- Tablets are an efficient way to add binder to XRF sample disks.
- Liquid easily mixes with the sample.
Chemical Identifiers
| SamplePrep Mixers, SamplePrep Mills | |
| Samples Which Crumble or Ablate After Pressing |
Specifications
| SamplePrep Mixers, SamplePrep Mills | |
| Samples Which Crumble or Ablate After Pressing | |
| 150 g |
| Paraffin Binder | |
| ≤20 μm |