Learn More
Cole-Parmer™ XRF Prep-Aid™ Grinding Aids and Sample Binders
Prepare samples for reproducible and contamination-free results with XRF Prep-Aid™ Grinding Aids and Sample Binders.
Supplier:  Cole-Parmer™ 3642 450
 View more versions of this product
Catalog No. 3642450
Only null left
Description
Description
Ideal for samples which crumble or ablate after pressing.
  • Select binders from powder, tablet, or liquid form depending on your application.
  • High-quality sample preparation is the key to achieving consistent and reliable analytical results.
  • Powders are ideal for samples that crumble or ablate after pressing.
  • They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks.
  • Tablets are an efficient way to add binder to XRF sample disks.
  • Liquid easily mixes with the sample.
Specifications
Chemical Identifiers
SamplePrep Mixers, SamplePrep Mills
Samples Which Crumble or Ablate After Pressing
Specifications
SamplePrep Mixers, SamplePrep Mills
Samples Which Crumble or Ablate After Pressing
450 g
Cellulose Binder
≤20 μm
Product Suggestions
Product Suggestions