Learn More
Cole-Parmer™ XRF Prep-Aid™ Grinding Aids and Sample Binders
Catalog No. 3642150
Change view
Click to view available options
Type:
Cellulose Binder
Paraffin Binder
Ultrabind
Quantity:
150 g
450 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
3642150 Cellulose Binder 150 g
3642450 Cellulose Binder 450 g
3644450 Ultrabind 450 g
3646150 Paraffin Binder 150 g
3646450 Paraffin Binder 450 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 3642150 Supplier Cole-Parmer™ Supplier No. 3642 150
Prepare samples for reproducible and contamination-free results with XRF Prep-Aid™ Grinding Aids and Sample Binders.
Ideal for samples which crumble or ablate after pressing.
  • Select binders from powder, tablet, or liquid form depending on your application.
  • High-quality sample preparation is the key to achieving consistent and reliable analytical results.
  • Powders are ideal for samples that crumble or ablate after pressing.
  • They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks.
  • Tablets are an efficient way to add binder to XRF sample disks.
  • Liquid easily mixes with the sample.
Specifications
For Use With (Equipment) SamplePrep Mixers, SamplePrep Mills
Type Cellulose Binder
For Use With (Application) Samples Which Crumble or Ablate After Pressing
Particle Size ≤20 μm
Quantity 150 g