Learn More
Cole-Parmer™ Rack for Small Grinding Vials
Catalog No. 6755
Change view
Click to view available options
Holds:
Vials
Catalog No. Holds
6755 Vials
1 options
Catalog No. 6755 Supplier Cole-Parmer™ Supplier No. 6755
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Rack for Small Grinding Vials for Freezer/Mill™ cryogenic grinders to simplify your processes.
  • Closed grinding vial prevents cross-contamination.
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles.
  • Holds small grinding vial set, small poly-vial set, small Cr-free grinding vial set.
  • Compatible with Freezer/Mill™ cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
  • Durable glass-reinforced acetal construction
  • Volume: 0.5 to 4 mL
Specifications
Holds Vials
Material Glass-reinforced Acetal
Positions 16
Shape Rectangular, Rounded Corners
For Use With (Equipment) Small Grinding Vials
Product Type Vial Rack