missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Disposable Closed XRF X-Cell Sample Cups

Catalog No. p-61626
Change view
Click to view available options
Diameter (English):
1.2 in.
1.57 in.
1.7 in.
Diameter (Metric):
31 mm (micro)
31 mm
40 mm
43 mm
Top Style:
Closed
Closed, Collar
Double Open Ended, Collar
Open, Collar
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Diameter (Metric) Top Style
0467511 1.57 in. 40 mm Closed
0467512 1.2 in. 31 mm Closed
0467513 1.2 in. 31 mm Closed, Collar
0467514 1.7 in. 43 mm Closed
0467515 1.2 in. 31 mm Double Open Ended, Collar
0467516 1.2 in. 31 mm (micro) Open, Collar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 0467511 Supplier Cole-Parmer™ Supplier No. SPX3527
Only null left

Requires

Window film

Use these Disposable Closed XRF X-Cell Sample Cups for fast, dependable, and reproducible XRF analysis of liquid samples.

  • One opening for fast liquid sample delivery.
  • Compatible with major suppliers of XRF spectrometers.
  • Available in 31 mm, 40 mm, and 43 mm outer diameters.
  • Contaminant-free polyethylene construction.
  • Collars available to attach window film to cup for added security.

Cole-Parmer™ X-Cells are used throughout the world for fast, dependable, reproducible running of liquid samples in XRF spectrometers.

  • They are designed to fit into the major suppliers of XRF spectrophotometers, such as PANalytical, Rigaku, Bruker, and Thermo Instruments.
  • The durable polyethylene construction is generally resistant to acids, bases, and most organic chemicals except halogenated hydrocarbons.
  • The patented closed X-Cell design features a snap-open seal for venting volatile samples and a reservoir cup for catching any liquids that expands through the vent.
  • This assures the analyst of a smooth, fat window for improved precision, yet guards against spillage in inverted-optic spectrometers.
  • A roughed surface within the reservoir cup facilitates labeling.
  • Each closed X-Cell consists of a cup and snap-on ring.
  • For routine liquid samples, the cup is filled with liquid, a piece of film is spready over the top, then the snap-ring is pushed down over the cup to form a fat drum-like window.
    • The cell is turned over and, if desired, the snap-post on the rear can be pushed to break the seal and to permit venting of trapped bubbles and expand liquid while the sample is run.
    • The filled X-Cell is then positioned in the spectrometer mask.
  • Use the bubble-free insert with the 3527 (04576-05) 40 mm XRF liquid cell to prevent trapped air from forming a bubble under the window film when the cell is upright.
    • The insert is pushed into the cell before the sample is added and forms an inverted-cone shelf under which air bubbles can be trapped once the cell is filled and sealed.

Order Info

Special application versions including the 31 mm micro X-Cell—with a 6.3 mm centered window for small, hazardous, or air-sensitive samples—and the 43 mm cell for Horiba™ sulfur analyzers also available. Supplied in bags of 100 cups.

Specifications

For Use With (Equipment) XRF Spectrometers
Type Sample Cup
Diameter (English) 1.57 in.
Diameter (Metric) 40 mm
For Use With (Application) XRF Analysis
Material Polyethylene
Quantity 100 Pk.
Top Style Closed