missing translation for 'onlineSavingsMsg'
Learn More

Spermine tetrahydrochloride, 99%

Catalog No. AAJ6306003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6306003 1 g
AAJ6306006 5 g
AAJ6306014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6306003 Supplier Thermo Scientific Chemicals Supplier No. J6306003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Binds to the polyamine modulatory site of NMDA | CAS: 306-67-2 | C10H30Cl4N4 | 348.18 g/mol

Spermine tetrahydrochloride, is used to precipitate DNA from low salt aqueous buffers. Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors2 and red blood cells of chicken.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Spermine tetrahydrochloride, is used to precipitate DNA from low salt aqueous buffers. Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors2 and red blood cells of chicken.

Solubility
Soluble in water (348.2 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Store under inert gas. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 306-67-2
Molecular Formula C10H30Cl4N4
Molecular Weight (g/mol) 348.18
MDL Number MFCD00012914
InChI Key XLDKUDAXZWHPFH-UHFFFAOYSA-N
Synonym spermine tetrahydrochloride, n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride, gerontine tetrahydrochloride, neuridine tetrahydrochloride, n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride, musculamine tetrahydrochloride, geontine tetrahydrochloride, spermine hcl, 1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride, spermine, tetrahydrochloride
PubChem CID 9384
IUPAC Name N,N'-bis(3-aminopropyl)butane-1,4-diamine;tetrahydrochloride
SMILES [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN

Specifications

Color White
Quantity 1 g
Beilstein 3911771
Solubility Information Soluble in water (348.2g/L at 20°C).
Formula Weight 348.19
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Spermine tetrahydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-189-8
RTECSNumber EJ7230000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only