missing translation for 'onlineSavingsMsg'
Learn More

Spermine, 97%

Catalog No. p-7047649
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1956203 1 g
AAL1956206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1956203 Supplier Thermo Scientific Chemicals Supplier No. L1956203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71-44-3 | C10H26N4 | 202.35 g/mol

Binds to the polyamine modulatory site of NMDA Spermine is essential for both normal and neoplastic tissue growth. It is involved in the modulation of calcium-dependent immune processes. It plays an important role in cellular proliferation and differentiation as well as inhibits neuronal nitric oxide synthase (nNOS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Binds to the polyamine modulatory site of NMDA Spermine is essential for both normal and neoplastic tissue growth. It is involved in the modulation of calcium-dependent immune processes. It plays an important role in cellular proliferation and differentiation as well as inhibits neuronal nitric oxide synthase (nNOS).

Solubility
Soluble in chloroform, methanol, water and lower alcohols. Insoluble in ether, benzene and petroleum ether.

Notes
Air sensitive and hygroscopic. Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.

Chemical Identifiers

CAS 71-44-3
Molecular Formula C10H26N4
Molecular Weight (g/mol) 202.35
MDL Number MFCD00008215
InChI Key PFNFFQXMRSDOHW-UHFFFAOYSA-N
Synonym spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl
PubChem CID 1103
ChEBI CHEBI:15746
IUPAC Name N,N'-bis(3-aminopropyl)butane-1,4-diamine
SMILES NCCCNCCCCNCCCN

Specifications

Melting Point 28°C to 30°C
Boiling Point 150°C (5 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3259
Beilstein 1751791
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,8743
Solubility Information Soluble in chloroform,methanol,water and lower alcohols. Insoluble in ether,benzene and petroleum ether.
Formula Weight 202.35
Percent Purity 97%
Chemical Name or Material Spermine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 200-754-2
RTECSNumber EJ7175000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only