Learn More
3M™ Speedglas™ Outside Protection Plate
Catalog No. 296368091
Change view
Click to view available options
Replacement Parts & Accessories
Catalog No.
296368091 Replacement Parts & Accessories
1 options
Catalog No. 296368091 Supplier 3M™ Supplier No. 7010341129
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed to help protect the filter from spark and spatter. Failure to use an outside protection plate with the filter may void the warranty if spark or spatter contaminates the glass. 9100, 06-0200-52-B
  • Replacement part intended for use with 3M™ Speedglas™ Welding Helmet Series 9100.
  • The helmets come in a variety of shapes and sizes
  • They help to protect your eyes and face from radiation, heat and sparks, while providing a precise view of your work
  • 3M™ Speedglas™ Welding Helmet is sold separately.
Specifications
Product Type Replacement Parts and Accessories