Learn More
3M™ Speedglas™ Magnification Plate
Catalog No. 296368097
Change view
Click to view available options
Power Intensity:
1.5 x magnification
2.5 x magnification
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Power Intensity
296368097 2.5 x magnification
296368095 1.5 x magnification
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 296368097 Supplier 3M™ Supplier No. 7000127145
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The 3M™ Speedglas™ Magnification Plate (9100 Series) is used for magnifying objects
  • 3M™ Speedglas™ Magnification Plate can be inserted into the 3M™ Speedglas™ Auto Darkening Filter on the inside slot
Specifications
For Use With (Application) Assembly, Facility Maintenance, Grinding, Machine Operations, Sanding, Welding
Power Intensity 2.5 x magnification
Product Type Replacement Parts and Accessories