missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Spectrum Tubes
For demonstrating spectral lines of pure substances.
Supplier: Eisco™ PH1196B7
Includes:
Spectrum Tube
Description
- Features large 10cm capillary in center for greater viewing area
- For use with power supplies holding 26cm spectrum tubes
- Designed for operation intervals of 30 seconds
- Should be used for 30 seconds on/30 seconds off
For intermittent use only, not to be used as a lamp source.
Specifications
| Spectrum Tube, Hydrogen | |
| Demonstrates the spectral lines of pure substances | |
| Spectrum Tube, large 10cm capillary |