missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Spectrum Tubes

For demonstrating spectral lines of pure substances.

Supplier:  Eisco™ PH1196B11

 View more versions of this product

Catalog No. S13006


Only null left
Explore available promotions

Includes:

Spectrum Tube


Description

Description

  • Features large 10cm capillary in center for greater viewing area
  • For use with power supplies holding 26cm spectrum tubes
  • Designed for operation intervals of 30 seconds
  • Should be used for 30 seconds on/30 seconds off

For intermittent use only, not to be used as a lamp source.

Specifications

Specifications

Krypton Spectrum Tube
Demonstrates the spectral lines of pure substances
Spectrum Tube, large 10cm capillary
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions