missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO Spectrophotometer ScanStart™ Easy UV Water Testing Analyzer
Description
- ScanStart™ bundles equipped with barcode scanners enable seamless access to over 80 common wastewater, drinking water, and process water analytes and support flexible operation with NANOCOLOR™ test kits from Macherey-Nagel
- NANOCOLOR™ test kits greatly simplify sample preparation by including all necessary reagents, effectively eliminating the need for time-consuming and costly chemical preparation steps.
- Designed for simplicity and ease of use, delivering highly accurate and consistent measurement results, as well as built-in validated methods for up to 30 color scales that automatically eliminate calculation and transcription errors.
- Optical design is optimized to reduce effects of turbidity commonly found in wastewater samples
- Avoid errors. PathDetect™ securely checks the path length to ensure accurate results
- Enhanced usability features like a simplified display of concentration results and embedded performance tests significantly accelerate routine analyses.
- EasyDirect Software enables data acquisition and management as well as export capability for ease of result access and review. An intuitive interface displays concentration results of pertinent analytes for EPA methodologies in water analysis
Specifications
Specifications
| Detector Type | Dual Silicon Photodiodes |
| Display | Touchscreen |
| Languages | English, French, German, Chinese, Italian |
| Light Source | Xenon Flash |
| Spectral Bandwidth | 3 nm |
| Type | UV Spectrophotometer |
| Voltage | 100/240 V |
| Wavelength Range | 190 to 1000 nm |