missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Spectrophotometer Cuvettes

Catalog No. 111011703
Change view
Click to view available options
Capacity (Metric):
≥1 mL
≥2.5 mL
≥3 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
111011703 ≥1 mL
111011702 ≥2.5 mL
111011396 ≥3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111011703 Supplier MilliporeSigma™ Supelco™ Supplier No. C5416100EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Packaged in styrofoam boxes to assure cuvettes are dust- and scratch-free

  • These cuvettes are incompatible with some organic solvents

Specifications

Type Spectrophotometer Cuvette
Description Without Stopper
Capacity (Metric) ≥1 mL
Material Polystyrene
Quantity 100/Pk.
Path Length 10 mm
Wavelength Range 340 to 800 nm
For Use With (Application) UV/Vis Spectroscopy