Learn More
Thermo Scientific™ SPECTRONIC™ 200 Spectrophotometer Accessories
Catalog No. 361044430
Change view
Click to view available options
Type:
Cell Holder
Cuvette Rack
Replacement Sample Compartment
SPECTRONIC Standards 2 Kit
Thermal Printer
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
361044430 Thermal Printer
14385495 Cuvette Rack
361031141 Cell Holder
13875126 Replacement Sample Compartment
14380008 SPECTRONIC Standards 2 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 361044430 Supplier Thermo Scientific™ Supplier No. 840234000
Accessories for Thermo Scientific™ SPECTRONIC 200 Spectrophotometer
The SPECTRONIC 200 is a rugged, simple-to-use spectrophotometer ideal for routine applications. On-board software, controlled from an intuitive navigation pad using a full color screen, delivers capabilities beyond expectations.
Rugged, Practical Design
  • With no internal parts, there's nothing to wear out
  • Sample compartment lifts out for easy cleaning
  • Polymer construction resists corrosion from acidic or oxidizing solutions
  • Navigation pad and on-screen menus are second nature to any modern user
  • Compact footprint saves space in your lab
  • Cuvette racks on both sides of the sample compartment help you keep track of samples
  • No computer or serial cable required; Just plug it in and you're ready to measure
The Perfect Tool for Routine Spectroscopic Measurements
  • Large, bright, color LCD adjusts to the perfect angle for any technician to view
  • Standard sample compartment accommodates both 10mm square cuvettes and test tubes up to 13mm (0.5 inch) diameter
  • Room light immune optical system lets you measure tall test tubes with the lid open
  • Analyzer mode allows you to program standard methods
  • Screen-print feature gives a permanent record of your data
  • Quantitative analysis with either a factor or up to four standards offers confidence
  • Available Thermo Scientific VISIONlite5 software, for your computer, controls the instrument and provides data manipulation, storage and report printing options
Not ready to part with your old Thermo Scientific Spec 20 or GENESYS 20
  • The SPECTRONIC 200 will emulate the interface and controls on your old instrument to integrate seamlessly into your laboratory
Check the performance and accuracy of your spectrophotometer right in your own laboratory using the Thermo Scientific™ SPECTRONIC™ Spectrophotometer Standards. This kit is as easy to use as a standard cuvette and contains:
  • Calibrated neutral density glass standards for checking photometric accuracy at nominal values of 0.3A, 0.5A, 1A and 1.5A
  • Calibrated standard for checking wavelength accuracy at nominal wavelengths on 400nm, 522nm and 777nm
  • Filter for checking stray light in UV-Vis instruments at 220nm
  • Filter for checking stray light in Vis instruments at 400nm
  • Set of 7 glass pieces mounted in metal frames sized to fit standard square cuvette holders
Four calibrated neutral density glass standards for checking photometric accuracy at nominal values of 0.3A, 0.5A, 1A and 1.5A Standards are calibrated at 440.0n, 465.0nm, 546.1nm, 590.0nm and 635.0nm
One calibrated standard for checking wavelength accuracy at nominal wavelengths on 400nm, 522nm and 777nm
The symmetry of the peaks means that tests of wavelength accuracy are independent of the spectral bandwidth (between 1nm and 10nm) of the instrument being tested
1 filter for checking stray light in UV-Vis instruments at 220nm
1 filter for checking stray light in Vis instruments at 400nm
Each set of SPECTRONIC Standards is individually tested and certified to be accurate, with values traceable to the National Institute of Standards and Technology (NIST) or other Primary Standards
Filters are supplied in a tough plastic storage container
Recalibration service is available from Thermo Fisher Scientific. Recommended recalibration interval is two years
Note: Stray light filters should block light completely at the measurement wavelength and are therefore not something that can be calibrated.
Specifications
Type Thermal Printer
For Use With (Equipment) SPECTRONIC 200/200E Spectrophotometers