Learn More
Thermo Scientific™ SPECTRONIC™ 200 Spectrophotometer Accessories
Catalog No. 13875126
Change view
Click to view available options
Type:
Cell Holder
Cuvette Rack
Replacement Sample Compartment
SPECTRONIC Standards 2 Kit
Thermal Printer
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13875126 Replacement Sample Compartment
14385495 Cuvette Rack
361031141 Cell Holder
14380008 SPECTRONIC Standards 2 Kit
361044430 Thermal Printer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13875126 Supplier Thermo Scientific™ Supplier No. 840250700
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Accessories for Thermo Scientific™ SPECTRONIC 200 Spectrophotometer
The SPECTRONIC 200 is a rugged, simple-to-use spectrophotometer ideal for routine applications. On-board software, controlled from an intuitive navigation pad using a full color screen, delivers capabilities beyond expectations.
Rugged, Practical Design
  • With no internal parts, there's nothing to wear out
  • Sample compartment lifts out for easy cleaning
  • Polymer construction resists corrosion from acidic or oxidizing solutions
  • Navigation pad and on-screen menus are second nature to any modern user
  • Compact footprint saves space in your lab
  • Cuvette racks on both sides of the sample compartment help you keep track of samples
  • No computer or serial cable required; Just plug it in and you're ready to measure
The Perfect Tool for Routine Spectroscopic Measurements
  • Large, bright, color LCD adjusts to the perfect angle for any technician to view
  • Standard sample compartment accommodates both 10mm square cuvettes and test tubes up to 25mm (1 inch) diameter
  • Room light immune optical system lets you measure tall test tubes with the lid open
  • Analyzer mode allows you to program standard methods
  • Screen-print feature gives a permanent record of your data
  • Quantitative analysis with either a factor or up to four standards offers confidence
  • Available Thermo Scientific VISIONlite5 software, for your computer, controls the instrument and provides data manipulation, storage and report printing options
Not ready to part with your old Thermo Scientific Spec 20 or GENESYS 20
  • The SPECTRONIC 200 will emulate the interface and controls on your old instrument to integrate seamlessly into your laboratory
VISIONlite5 software — Volume pricing begins at eight copies and is available on request.
Specifications
Type Replacement Sample Compartment
For Use With (Equipment) SPECTRONIC 200/200E Spectrophotometers
Holds Test tube and 10mm cuvette