missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Spectral Calibration Plate with Alexa 647 Dye, 96-well

Catalog No. A50551
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50551 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A50551 Supplier Thermo Scientific™ Supplier No. A50551

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The 96-well Spectral Calibration Plate with Alexa Fluor 647 is used to maintain the QuantStudio Dx, QuantStudio 5 Dx, and QuantStudio 7 Pro Dx real-time PCR instruments with 96-well blocks.

The 96-well Spectral Calibration Plate with Alexa Fluor 647 is used to maintain the QuantStudio Dx, QuantStudio 5 Dx, and QuantStudio 7 Pro Dx real-time PCR instruments with 96-well blocks. Please see the instrument user guide for full instructions on use.

Specifications

Format 96-well, 0.2 mL
Content And Storage Store at -20°C ± 5°C.
Calibrated Dye AlexaFluor™ 647
For Use With (Equipment) QuantStudio™ 5 Dx, QuantStudio™ 7 Pro Dx
Product Type Calibration Plate
For Use With (Application) Spectral Calibration
Shipping Condition Dry Ice
Quantity 1 Plate
Calibration Type Spectral Calibration

For In Vitro Diagnostic Use.