missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin dihydrochloride pentahydrate, 50 mg/ml in water

Catalog No. AAJ67387XF
Change view
Click to view available options
Quantity:
1 mL
5 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67387XF 1 mL
AAJ673878EQ 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67387XF Supplier Thermo Scientific Chemicals Supplier No. J67387XF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22189-32-8 | C14H36Cl2N2O12 | 495.34 g/mol

It has been shown to have a wide variety of uses, including treating acute gonorrheal urethritis and cervictis, to mark cell layers to monitor cell fate during leaf development, as a selection marker in plant related transformation systems for plant cells containing the marker gene Spcr, to study respiratory tract infections of cattle caused by Pasteurella multocida and Mannheimia haemolytica, and to generate plants deficient for the plastid-encoded RNA polymerase on MS-agar media. Spectinomycin also has use in amplification of low copy number plasmid carrying replicons. It is recommended for use in cell culture applications at 7.5-20 mg/L.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been shown to have a wide variety of uses, including treating acute gonorrheal urethritis and cervictis, to mark cell layers to monitor cell fate during leaf development, as a selection marker in plant related transformation

Chemical Identifiers

CAS 22189-32-8
Molecular Formula C14H36Cl2N2O12
Molecular Weight (g/mol) 495.34
MDL Number MFCD00150886
InChI Key DCHJOVNPPSBWHK-UXXUFHFZSA-N
PubChem CID 67108398
ChEBI CHEBI:9217
IUPAC Name dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride
SMILES [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O

Specifications

Type Spectinomycin Dihydrochloride Pentahydrate
Physical Form Liquid
Packaging Plastic Bottle
Quantity 1 mL
Solubility Information Fully miscible in water. Soluble in methanol,DMSO (∽11mg/ml),ethanol (∽0.3mg/ml),and DMF (∽2mg/ml).
Chemical Name or Material Spectinomycin dihydrochloride pentahydrate
Hazard Category H335
EINECSNumber 244-554-3
RTECSNumber WG7400000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only