missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectinomycin dihydrochloride, 96.4%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0215206705
| Quantity | 5 g |
|---|
Chemical Identifiers
| 22189-32-8 | |
| 495.34 | |
| DCHJOVNPPSBWHK-UXXUFHFZSA-N | |
| 67108398 | |
| dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride |
| C14H36Cl2N2O12 | |
| MFCD00150886 | |
| U18409AE, Actinospectacin | |
| CHEBI:9217 | |
| [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O |
Specifications
| Spectinomycin Dihydrochloride | |
| Crystalline Powder | |
| 5 g | |
| C14H36Cl2N2O12 | |
| U18409AE, Actinospectacin | |
| [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O | |
| 495.34 | |
| CHEBI:9217 |
| Spectinomycin dihydrochloride | |
| 22189-32-8 | |
| 0.964 | |
| MFCD00150886 | |
| DCHJOVNPPSBWHK-UXXUFHFZSA-N | |
| dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride | |
| 67108398 | |
| 495.36 |