missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin dihydrochloride, 96.4%, MP Biomedicals™

Catalog No. ICN15206701
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15206701 1 g
ICN15206705 5 g
ICN15206725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15206701 Supplier MP Biomedicals Inc Supplier No. 0215206701

Chemical Identifiers

CAS 22189-32-8
Molecular Formula C14H36Cl2N2O12
Molecular Weight (g/mol) 495.34
MDL Number MFCD00150886
InChI Key DCHJOVNPPSBWHK-UXXUFHFZSA-N
Synonym U18409AE, Actinospectacin
PubChem CID 67108398
ChEBI CHEBI:9217
IUPAC Name dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride
SMILES [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O

Specifications

Type Spectinomycin Dihydrochloride
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 0.964
Formula Weight 495.36
Chemical Name or Material Spectinomycin dihydrochloride