missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Specimen Temperature Control

Catalog No. 01672881
Change view
Click to view available options
Conjugate:
None
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
01672881 None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01672881 Supplier BINDER™ Supplier No. 80122343

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specimen temperature control with additional specimen temperature sensor.

  • Sensor with steel-covered, heat-resistant line
  • Additional indication of the specimen temperature on the display

Specifications

Applications Not Available
Conjugate None
Description Specimen Temperature Control
For Use With (Equipment) Units Series M