missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Specimen Chucks for QS and Microm Cryostats, 40 mm

Catalog No. 22445881
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Product Type:
Circular Chuck
Waffle Chuck
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
22445881 White Circular Chuck
22445745 Blue Circular Chuck
22445836 Green Circular Chuck
22445259 Orange Circular Chuck
22445293 Pink Circular Chuck
22445638 Purple Circular Chuck
22445494 Red Circular Chuck
22445663 Yellow Circular Chuck
22445498 Blue Waffle Chuck
22445721 Green Waffle Chuck
22445483 Orange Waffle Chuck
22445484 Pink Waffle Chuck
22445661 Purple Waffle Chuck
22445478 Red Waffle Chuck
22445481 White Waffle Chuck
22445499 Yellow Waffle Chuck
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 22445881 Supplier Mopec™ Supplier No. SP134WH

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specimen Chucks are available in multiple sizes and colors. Some chucks are also available in waffle pattern.

  • Round, aluminum specimen chucks

Specifications

Product Type Circular Chuck
For Use With (Equipment) QS and Microm Cryostats
Color White