missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Specimen Chucks for Leica CM1950 Cryostat, 40 mm

Catalog No. 22445791
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Product Type:
Circular Chuck
Waffle Chuck
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
22445791 Red Waffle Chuck
22445360 Blue Circular Chuck
22445396 Green Circular Chuck
22445398 Orange Circular Chuck
22445415 Pink Circular Chuck
22445447 Purple Circular Chuck
22445446 Red Circular Chuck
22445700 White Circular Chuck
22445703 Yellow Circular Chuck
22445448 Orange Waffle Chuck
22445733 Pink Waffle Chuck
22445735 Purple Waffle Chuck
22445704 White Waffle Chuck
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 22445791 Supplier Mopec™ Supplier No. SP114WRD

Specimen Chucks are available in multiple sizes and colors. Some chucks are also available in waffle pattern.

  • Round, aluminum specimen chucks

Specifications

Product Type Waffle Chuck
For Use With (Equipment) Leica CM1950 Cryostat
Color Red