missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mopec™ Specimen Chucks for Leica CM1950 Cryostat, 40 mm
Description
- Round, aluminum specimen chucks
Specifications
Specifications
| Product Type | Circular Chuck |
| For Use With (Equipment) | Leica CM1950 Cryostat |
| Color | Pink |