missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Specimen Chucks for Leica CM1950 Cryostat, 30 mm

Catalog No. p-200007557
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Product Type:
Circular Chuck
Waffle Chuck
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
22445507 Blue Circular Chuck
22445753 Green Circular Chuck
22445420 Orange Circular Chuck
22445754 Pink Circular Chuck
22445422 Purple Circular Chuck
22445647 Red Circular Chuck
22445580 White Circular Chuck
22445679 Yellow Circular Chuck
22445811 Blue Waffle Chuck
22445878 Green Waffle Chuck
22445401 Orange Waffle Chuck
22445876 Pink Waffle Chuck
22445402 Purple Waffle Chuck
22445812 Red Waffle Chuck
22445261 White Waffle Chuck
22445399 Yellow Waffle Chuck
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 22445507 Supplier Mopec™ Supplier No. SP113BL
Only null left

Specimen Chucks are available in multiple sizes and colors. Some chucks are also available in waffle pattern.

  • Round, aluminum specimen chucks

Specifications

Product Type Circular Chuck
For Use With (Equipment) Leica CM1950 Cryostat
Color Blue