missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Specimen Chucks for CRYOSTAR Cryostats, 30 mm

Catalog No. p-200007551
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Product Type:
Circular Chuck
Waffle Chuck
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
22445510 Blue Circular Chuck
22445646 Green Circular Chuck
22445744 Orange Circular Chuck
22445623 Pink Circular Chuck
22445325 Purple Circular Chuck
22445326 Red Circular Chuck
22445309 White Circular Chuck
22445323 Yellow Circular Chuck
22445882 Blue Waffle Chuck
22445628 Green Waffle Chuck
22445304 Orange Waffle Chuck
22445695 Pink Waffle Chuck
22445662 Purple Waffle Chuck
22445332 Red Waffle Chuck
22445719 White Waffle Chuck
22445657 Yellow Waffle Chuck
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 22445510 Supplier Mopec™ Supplier No. SP123BL
Only null left

Specimen Chucks are available in multiple sizes and colors. Some chucks are also available in waffle pattern.

  • Round, aluminum specimen chucks

Specifications

Product Type Circular Chuck
For Use With (Equipment) CryoStar NX50 and NX70
Color Blue