missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Specific Gravity Metal Blocks

Catalog No. S12917
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S12917 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S12917 Supplier Eisco™ Supplier No. PH0111

Use the Specific Gravity Metal Blocks for the study of gravity, density, and mass.

  • Set of four metal blocks
  • Aluminum, brass, copper, steel
  • Instruction manual provides a background on density, specific gravity and specific heat and includes inquiry based student activities with capture sheet
  • Helps students identify and figure out mass; understand the relationship between mass, volume and density; and understand specific heat and specific gravity

Specifications

Grade 7 to 12
For Use With (Application) For the study of gravity, density, and mass.
Includes Set of four metal blocks
Material Aluminum, Brass, Copper, Steel
Dimensions (L x W x H) 50 x 13 x 13 mm
Class Size 1 Lab Group
Product Type Specific Gravity Metal Blocks