missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Spec-Tec™ Sterile Cut Resistant Glove Liners - ANSI A2

Catalog No. 19169810A
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19169810A Small
19169810B Medium
19169810C Large
19169810D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19169810A Supplier Wells Lamont™ Supplier No. M104S
Only null left

HPPE fibers are comfortable and stretchy, while also providing ANSI Cut Level A2 protection. Each pair of Spec-Tec™ Sterile Cut Resistant Glove Liners is individually wrapped around tissue paper in a secure, sterile package.

Lightweight and comfortable

  • Sterile liners are constructed from a blend of Lycra™ and HPPE fibers
  • Liners provide ANSI Cut Level A2 protection
  • Individually wrapped around tissue paper in secure, sterile packaging
  • Single use in sterile environments
  • Launderable for reuse in non-sterile environments
  • Latex-free
  • Made in the USA

Specifications

Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A2
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material HPPE, Lycra
Product Type Cut-resistant Glove Liners
Size Small
Sterility Sterile
For Use With (Application) Medical, Organ Tissue Processing, and Pharmaceuticals
Glove Cuff Style Knit
Hem Color Green
Inspected for Medical Use Yes
Packaging Type Individual
Series M104
Show More Show Less