missing translation for 'onlineSavingsMsg'
Learn More

Spearmint Oil, Natural, FCC, 55%, Spectrum™ Chemical

SP306, 8008-79-5

$368.86 - $83260.43

Chemical Identifiers

CAS 8008-79-5
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606968
View Documents
Spectrum Chemical Mfg Cor
SP306100ML
100 mL Amber Glass Bottle
Each for $368.86
Only null left
 
18606969
View Documents
Spectrum Chemical Mfg Cor
SP306500MLGL
500 mL Amber Glass Bottle
Each for $1,159.00
Only null left
 
18606970
View Documents
Spectrum Chemical Mfg Cor
SP30625LTGL
2.5 L Amber Glass Bottle
Each for $3,161.74
Only null left
 
18606972
View Documents
Spectrum Chemical Mfg Cor
SP306200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18606971
View Documents
Spectrum Chemical Mfg Cor
SP30625LBBL
25 lb. Steel Pail
Each for $7,958.29
Only null left
 
18606973
View Documents
Spectrum Chemical Mfg Cor
SP30650KGBL
50 kg Steel Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Spearmint Oil, Natural, FCC is used for cooking, flavoring, and as a digestive support. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

8008-79-5

Specifications

8008-79-5
Amber Glass Bottle
100 mL
FCC
1
1.484-1.491
55%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'